Läs mer
Methylpropylnitrosamine, TRC
Produktkod. 30479910
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30479910 250 mg
1 options
Produktkod. 30479910 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM327205250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methylpropylnitrosamine
CAS 924-46-9
Anteckningar om renhetsgrad GC
Molekylformel C4 H10 N2 O
Kvantitet 250 mg
Synonym 1-Propanamine, N-methyl-N-nitroso- (9CI, ACI), N-Methyl-N-nitroso-1-propanamine (ACI), Propylamine, N-methyl-N-nitroso- (6CI, 7CI, 8CI), Methylpropylnitrosamine, N-Methyl-N-nitrosopropylamine, N-Nitroso-N-methyl-1-propanamine, N-Nitroso-N-methylpropylamine
InChI formel InChI=1S/C4H10N2O/c1-3-4-6(2)5-7/h3-4H2,1-2H3
LEDER CCCN(C)N=O
IUPAC-namn N-methyl-N-propyl-nitrous amide
Molekylvikt (g/mol) 102.14
Formel vikt 102.0793