Läs mer
Methylprotodioscin, TRC
Produktkod. 30475305
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30475305 50 mg
1 options
Produktkod. 30475305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32824550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methylprotodioscin
CAS 54522-52-0
Molekylformel C52H86O22
Kvantitet 50 mg
Synonym 22-Methoxylprotodioscin, 22-O-Methylprotodioscin, 3β,26-Dihydroxy-22α-methoxyfurost-5-ene 3-O-β-chacotrioside 26-O-β-D-glucopyranoside, Methylprotodioscin, NSC 698790, Smilax saponin B
InChI formel InChI=1S/C52H86O22/c1-21(20-66-46-40(61)39(60)36(57)31(18-53)70-46)10-15-52(65-7)22(2)33-30(74-52)17-29-27-9-8-25-16-26(11-13-50(25,5)28(27)12-14-51(29,33)6)69-49-45(73-48-42(63)38(59)35(56)24(4)68-48)43(64)44(32(19-54)71-49)72-47-41(62)37(58)34(55)23(3)67-47/h8,21-24,26-49,53-64H,9-20H2,1-7H3/t21-,22+,23+,24+,26+,27-,28+,29+,30+,31-,32-,33+,34+,35+,36-,37-,38-,39+,40-,41-,42-,43+,44-,45-,46-,47+,48+,49-,50+,51+,52-/m1/s1
LEDER CCOC(=O)C(\C)=C/C[C@@H](/C(C)=C/c1nc(C)sc1)O[Si](C)(C)C(C)(C)C
Molekylvikt (g/mol) 1063.23