Läs mer
Metiprenaline, TRC
Produktkod. 30478622
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30478622 100 mg
1 options
Produktkod. 30478622 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM338645100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Metiprenaline
CAS 1212-03-9
Molekylformel C12 H19 N O3
Kvantitet 100 mg
Synonym 4-Hydroxy-3-methoxy-α-[[(1-methylethyl)amino]methyl]-benzenemethanol, α-[(Isopropylamino)methyl]-vanillyl Alcohol, 4-Hydroxy-3-methoxy-α-[[(1-methylethyl)amino]methyl]benzenemethanol, 3-Methoxyisoprenaline, 3-Methoxyisoproterenol, 3-O-Methylisoprenaline, 3-O-Methylisoproterenol, Methylprenalin, Methylprenaline, Moxa, Moxa (pharmaceutical), (1-Methylethyl)-normetadrenaline, O-Methylisoprenaline
InChI formel InChI=1S/C12H19NO3/c1-8(2)13-7-11(15)9-4-5-10(14)12(6-9)16-3/h4-6,8,11,13-15H,7H2,1-3H3
LEDER COc1cc(ccc1O)C(O)CNC(C)C
IUPAC-namn 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]-2-methoxyphenol
Molekylvikt (g/mol) 225.284
Formel vikt 225.136