Learn More
Metixene EP Impurity B, Mikromol™
Product Code. 30050127
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050127 25 mg 25mg
30050905 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30050127 Supplier Mikromol Supplier No. MM0125.020025
 Restricted Item
Beräknad leverans: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Metixene EP Impurity B
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Thioxanthone
CAS 492-22-8
Analyte or Component Names Thioxanthone
Shipping Condition Room Temperature
Molecular Formula C13 H8 O S
Certifications/Compliance ISO 17025
Synonym 9H-Thioxanthen-9-one (Thioxanthone), 9H-Thioxanthen-9-one, Metixene Hydrochloride Imp. B (EP), Thioxanthone, Metixene Hydrochloride Impurity B, Metixene Impurity B
InChI Formula InChI=1S/C13H8OS/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8H
SMILES O=C1c2ccccc2Sc3ccccc13
IUPAC Name thioxanthen-9-one
Molecular Weight (g/mol) 212.27
Quantity 25 mg
Formula Weight 212.0296 g/mol