Läs mer
Metixene hydrochloride hydrate, MedChemExpress
Produktkod. 16454908
Encompass_Preferred
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
16454908 10 mg
1 options
Produktkod. 16454908 Leverantör Medchem Express Leverantörsnummer HY120081A10mg
MedChemExpress Metixene hydrochloride hydrate is an anticholinergic antiparkinsonian agent, potently inhibits binding of quinuclidinyl benzilate (QNB) to the muscarinic receptor in rat brain cortical tissue, with an IC50 of 55 nM and a Kd of 15 nM.
Metixene hydrochloride hydrate is an anticholinergic antiparkinsonian agent, potently inhibits binding of quinuclidinyl benzilate (QNB) to the muscarinic receptor in rat brain cortical tissue, with an IC50 of 55 nM and a Kd of 15 nM.
Specifikationer
Kemiskt namn eller material Metixene hydrochloride hydrate
Fysisk form Solid
CAS 7081-40-5
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C20H26ClNOS
För användning med (applikation) Neuroscience-Neurodegeneration
Löslighetsinformation DMSO : 77.5 mg/mL (212.95 mM; Need ultrasonic) ∣H2O : 10 mg/mL (27.48 mM; Need ultrasonic)
LEDER CN1CC(CCC1)CC2C3=C(C=CC=C3)SC4=C2C=CC=C4.Cl.O
Molekylvikt (g/mol) 363.94
Formel vikt 363.94
Procent renhet 98.0%