Läs mer
Metoclopramide, TRC
Produktkod. 30008494
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008494 100 mg
30008460 500 mg
30009054 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM338698100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 364-62-5
Molekylformel C14 H22 Cl N3 O2
Molekylvikt (g/mol) 299.8
Synonym o-Anisamide, 4-amino-5-chloro-N-[2-(diethylamino)ethyl]- (7CI,8CI), 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide, 2-Methoxy-4-amino-5-chloro-N,N-dimethylaminoethylbenzamide, 2-Methoxy-5-chloroprocainamide, 4-Amino-5-chloro-2-methoxy-N-(β-diethylaminoethyl)benzamide, 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-o-anisamide, 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-o-anisamide, 5-Chloro-2-methoxyprocainamide, Afipran, Clopromate, DEL 1267, Draclamid, Emperal, Eucil, Gastrese, Gastro-Timelets, Gastro-tablinen, Gastrobid, Gastromax, Gastrosil, Gastrotem, MCP-ratiopharm, Macperan, Maxeran, Meclopran, Metamide, Methochlopramide, Metochlopramide, Metoclol, Metoclopramide, Metocobil, Metramid, Moriperan, N-(2-Diethylaminoethyl)-2-methoxy-4-amino-5-chlorobenzamide, N-(Diethylaminoethyl)-2-methoxy-4-amino-5-chlorbenzamide, N-(Diethylaminoethyl)-2-methoxy-4-amino-5-chlorobenzamide, N-[2-(Diethylamino)ethyl]-4-amino-5-chloro-2-methoxybenzamide, Orpamid, Parmid, Plasil, Plasil (pharmaceutical), Primperan, Regla, Reliveran
IUPAC-namn 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide
LEDER CCN(CC)CCNC(=O)c1cc(Cl)c(N)cc1OC
Specifikationer
Kemiskt namn eller material Metoclopramide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C14H22ClN3O2/c1-4-18(5-2)7-6-17-14(19)10-8-11(15)12(16)9-13(10)20-3/h8-9H,4-7,16H2,1-3H3,(H,17,19)
Formel vikt 299.1401
Procent renhet >95%