Läs mer
Metofenazate Fumarate, TRC
Produktkod. 30466082
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30466082 250 mg
1 options
Produktkod. 30466082 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM225805250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Metofenazate Fumarate
CAS 522-23-6
Molekylformel C31H36ClN3O5S • xC4H4O4
Kvantitet 250 mg
Synonym 3,4,5-Trimethoxybenzoic Acid 2-[4-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-1-piperazinyl]ethyl Ester Fumarate, 2-[4-[3-(2-Chlorophenothiazin-10-yl)propyl]-1-piperazinyl]ethyl 3,4,5-Trimethoxybenzoate, Methophenazine Fumarate
InChI formel InChI=1S/C31H36ClN3O5S.2C4H4O4/c1-37-26-19-22(20-27(38-2)30(26)39-3)31(36)40-18-17-34-15-13-33(14-16-34)11-6-12-35-24-7-4-5-8-28(24)41-29-10-9-23(32)21-25(29)35;2*5-3(6)1-2-4(7)8/h4-5,7-10,19-21H,6,11-18H2,1-3H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1+
LEDER OC(/C=C/C(O)=O)=O.ClC1=CC2=C(C=C1)SC3=CC=CC=C3N2CCCN4CCN(CCOC(C5=CC(OC)=C(OC)C(OC)=C5)=O)CC4
Molekylvikt (g/mol) 598.1511607