Läs mer
Metolachlor ESA Sodium Salt, TRC
Produktkod. 30010460
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010460 25 mg
17932225 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010460 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33871025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 947601-85-6
Molekylformel C15 H22 N O5 S . Na
Molekylvikt (g/mol) 351.39
Synonym CGA 354743, Metolachlor ethane sulfonic acid (ESA) sodium salt, Metolachlor-ethane sulfonic acid (ESA) sodium, Metolachlor ESA sodium salt, Ethanesulfonic acid, 2-[(2-ethyl-6-methylphenyl)(2-methoxy-1-methylethyl)amino]-2-oxo-, sodium salt (1:1) (ACI)
IUPAC-namn sodium;2-[2-ethyl-N-(1-methoxypropan-2-yl)-6-methylanilino]-2-oxoethanesulfonate
LEDER [Na+].CCc1cccc(C)c1N(C(C)COC)C(=O)CS(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Metolachlor ethane sulfonic acid (ESA) sodium salt
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C15H23NO5S.Na/c1-5-13-8-6-7-11(2)15(13)16(12(3)9-21-4)14(17)10-22(18,19)20;/h6-8,12H,5,9-10H2,1-4H3,(H,18,19,20);/q;+1/p-1
Formel vikt 351.1116
Procent renhet >95%