missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Metolachlor OA, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Metolachlor oxanilic acid (OA) |
| CAS | 152019-73-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C15 H21 N O4 |
| Kvantitet | 100 mg |
| Synonym | Metolachlor Oxanilic Acid (OA), Acetic acid, [(2-ethyl-6-methylphenyl)(2-methoxy-1-methylethyl)amino]oxo- (9CI), 2-[(2-Ethyl-6-methylphenyl)(2-methoxy-1-methylethyl)amino]-2-oxoacetic acid, CGA 51202, Metolachlor OA, Metolachlor OXA, Metolachlor oxanilic acid |
| InChI formel | InChI=1S/C15H21NO4/c1-5-12-8-6-7-10(2)13(12)16(11(3)9-20-4)14(17)15(18)19/h6-8,11H,5,9H2,1-4H3,(H,18,19) |
| LEDER | CCc1cccc(C)c1N(C(C)COC)C(=O)C(=O)O |
| IUPAC-namn | 2-[2-ethyl-N-(1-methoxypropan-2-yl)-6-methylanilino]-2-oxoacetic acid |
| Molekylvikt (g/mol) | 279.33 |
| Visa mer |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?