Läs mer
Metolachlor, TRC
Produktkod. 30480918
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30480918 100 mg
1 options
Produktkod. 30480918 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM338700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Metolachlor
CAS 51218-45-2
Molekylformel C15 H22 Cl N O2
Kvantitet 100 mg
Synonym 2-Chloro-N-(2-ethyl-6-methylphenyl)-N-(2-methoxy-1-methylethyl)acetamide, CGA 24705, Codal, Dual, Dual 720EC, Dual 960 EC, Dual II, Dual Magnum, Dual Triple, Jindual, Metetilachlor, Metoken, Metolachlor, N-(1-Methyl-2-methoxyethyl)-N-chloroacetyl-2-ethyl-6-methylaniline, Pennant, Yibingjiacaoan
InChI formel InChI=1S/C15H22ClNO2/c1-5-13-8-6-7-11(2)15(13)17(14(18)9-16)12(3)10-19-4/h6-8,12H,5,9-10H2,1-4H3
LEDER CCc1cccc(C)c1N(C(C)COC)C(=O)CCl
IUPAC-namn 2-chloro-N-(2-ethyl-6-methylphenyl)-N-(1-methoxypropan-2-yl)acetamide
Molekylvikt (g/mol) 283.79
Formel vikt 283.1339