Läs mer
Metomidate Hydrochloride, TRC
Produktkod. 30480466
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480466 25 mg
30472699 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30480466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33874525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35944-74-2
Molekylformel C13 H14 N2 O2 . Cl H
Molekylvikt (g/mol) 266.72
Synonym 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, methyl ester, hydrochloride (1:1), 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, methyl ester, monohydrochloride (9CI), Imidazole-5-carboxylic acid, 1-(α-methylbenzyl)-, methyl ester, hydrochloride (7CI), Metomidate hydrochloride, NSC 168403
IUPAC-namn methyl 3-(1-phenylethyl)imidazole-4-carboxylate;hydrochloride
LEDER Cl.COC(=O)c1cncn1C(C)c2ccccc2
Specifikationer
Kemiskt namn eller material Metomidate Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C13H14N2O2.ClH/c1-10(11-6-4-3-5-7-11)15-9-14-8-12(15)13(16)17-2;/h3-10H,1-2H3;1H
Formel vikt 266.0822
Procent renhet >95%