Läs mer
Metoprolol Acid Ethyl Ester, TRC
Produktkod. 30469665
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469665 100 mg
30464827 250 mg
30465328 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30469665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM338795100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29112-40-1
Molekylformel C16 H25 N O4
Molekylvikt (g/mol) 295.37
IUPAC-namn ethyl 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetate
LEDER CCOC(=O)Cc1ccc(OCC(O)CNC(C)C)cc1
Specifikationer
Kemiskt namn eller material Metoprolol Acid Ethyl Ester
Kvantitet 100 mg
InChI formel InChI=1S/C16H25NO4/c1-4-20-16(19)9-13-5-7-15(8-6-13)21-11-14(18)10-17-12(2)3/h5-8,12,14,17-18H,4,9-11H2,1-3H3
Formel vikt 295.1784