Läs mer
Metoprolol Acid, TRC
Produktkod. 30006667
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006667 50 mg
17901165 5 mg
17993635 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006667 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33878550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56392-14-4
Molekylformel C14 H21 N O4
Molekylvikt (g/mol) 267.32
Synonym 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetic Acid, Atenolol Imp. G (EP)
IUPAC-namn 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetic acid
LEDER CC(C)NCC(O)COc1ccc(CC(=O)O)cc1
Specifikationer
Kemiskt namn eller material 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C14H21NO4/c1-10(2)15-8-12(16)9-19-13-5-3-11(4-6-13)7-14(17)18/h3-6,10,12,15-16H,7-9H2,1-2H3,(H,17,18)
Formel vikt 267.1471
Procent renhet >95%