Learn More
SP Bel-Art™ Metric Color Micro Spinbar Magnetic Stirring Bars
Micro magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers.
Brand:  SP Bel-Art™ F37122-0303
Additional Details : Weight : 0.00570kg
 View more versions of this product
Product Code. 16845433
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Available in a variety of colors and sizes
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Specifications
Specifications
3 mm
Yellow
PTFE
3 mm
PTFE, FDA-grade, USP Class VI
10 Pk.