Läs mer
SP Bel-Art™ Metric Polygon Spinbar Magnetic Stirring Bars without Pivot Ring
Handla allt SP Scienceware Produkter
Längd (metrisk):
13 mm
16 mm
22 mm
25 mm
38 mm
50 mm
51 mm
60 mm
64 mm
70 mm
76 mm
80 mm
Diameter (metrisk):
7 mm
8 mm
10 mm
13 mm
Förpackningsstorlek:
Paket med 10
Paket med 5
Varje
Beskrivning
- Multi-faceted polygon shape with increased surface area creates added turbulence relative to similar sized, smooth cylindrical bars to provide thorough mixing
- Absence of a pivot ring allows the Spinbar to stay close to the bottom of the vessel enabling “scraping” of solids
- Optimal coupling with magnetic spindrive as entire length of stir bar is in close contact
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating; - Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Specifikationer
Specifikationer
| Autoklaveringsbar | Autoklaveringsbar |
| Färg | Vitt |
| Beläggning | PTFE, FDA-klassad, USP Klass VI |
| Diameter (metrisk) | 13 mm |
| Längd (metrisk) | 64 mm |
| Material | PTFE |