Learn More
Metro™ Stainless Laboratory Worktable Accessory, Drawer
Stow items away safely and neatly with a drawer add-on accessory. Metrol Stainless Laboratory Worktable Drawer comes in sizes for 30, 36, 48, or 60 in. (76, 91, 122, and 152cm) table lengths.
Brand:  Metro™ WTD21C
Additional Details : Weight : 2.54400kg
 View more versions of this product
Product Code. 15977341
  • 1900.00 SEK / Each
Estimated Shipment: 24-09-2024
to see stock.
Add to basket
Description
Description
  • Can be used with stationary or mobile worktablesl
Specifications
Specifications
14.25 in.
Economy Drawer
14.6 cm
5.75 in.
71.1 cm
28 in.
28 x 14.25 x 5.75 in. (71.1 x 36.1 x 14.6 cm)
36.1 cm
Stationary or mobile 30, 36, 48 and 60 in. W worktables
Smooth Chrome