Learn More
Metro™ Stainless Laboratory Worktable Accessory, Drawer
Stow items away safely and neatly with a drawer add-on accessory. Metrol Stainless Laboratory Worktable Drawer comes in sizes for 30, 36, 48, or 60 in. (76, 91, 122, and 152cm) table lengths.
Brand:  Metro™ WTD51S
Additional Details : Weight : 11.24800kg
 View more versions of this product
Product Code. 15987341
  • 10545.00 SEK / Each
Estimated Shipment: 24-09-2024
to see stock.
Add to basket
Description
Description
  • Can be used with stationary or mobile worktablesl
Specifications
Specifications
24 in.
Deluxe Drawer
19 cm
7.5 in.
64.6 cm
25.75 in.
25.75 x 24 x 7.5 in. (64.6 x 61 x 19 cm)
61 cm
Stationary or mobile 30, 36, 48 and 60 in. W worktables
Smooth Chrome