Learn More
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge Product Code.: 15946951
Estimated Shipment: 11-11-2025
Only null left
Add to basket
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge
Product Code. p-4247128
21 in., 4 in., Stainless Steel, Each
Click to view available options
Length (English):
14 in.
18 in.
21 in.
24 in.
30 in.
36 in.
42 in.
48 in.
60 in.
72 in.
Height (English):
1 in.
4 in.
Material:
Chrome
Stainless Steel
Unit Size:
Each
Product Code. 15946951
Brand: Metro™ L21WS
Only null left
Estimated Shipment: 11-11-2025
Add to basket
Only null left
Add to basket
Ledges are available for wire or solid shelves
  • 1 in. ledges provide a sturdy lip to secure items during transport or to meet seismic or other safety requirements.
  • Easily attach with no tools to Metro Super Erecta or Super Adjustable wire shelving.
  • Other sizes and finishes available.
  • For stationary or mobile installation.
  • Prevents items from protruding or falling from shelves.
  • Side and back ledges stand 1 in. (25 mm) high.
  • Solid shelf ledges stand 4 in. high (10.1 cm) and include spring-clip tabs for attaching.
Order Info
Posts and shelves sold separately.
Specifications
Product Type Stackable Ledge for Solid Shelving
Material Stainless Steel
For Use With (Equipment) Metro Super Erecta Solid Shelves
Height (English) 4 in.
Height (Metric) 10 cm
Includes Spring clip tabs
Length (English) 21 in.
Length (Metric) 53.3 cm
Product Content Correction
Product Title
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge >