Learn More
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge, Stackable
Brand:  Metro™ L72N4C
Additional Details : Weight : 2.27000kg
Product Code. 11775699
  • 610.00 SEK / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
  • 4 in. high (10cm)
  • Choice of plated or stainless steel finish
Order Info
Posts and shelves sold separately.
Specifications
Specifications
72 in.
182.8 cm
Metro™ Super Erecta™ wire shelving
10 cm
4 in.
Chrome