Learn More
Metro™ Wire Shelving Track Sets Product Code.: 15957311
Only undefined left
Add to basket
Metro™ Wire Shelving Track Sets
Product Code. 15957311
204 in., Each
Click to view available options
Length (English):
27 in.
72 in.
84 in.
96 in.
108 in.
120 in.
132 in.
144 in.
156 in.
168 in.
180 in.
192 in.
204 in.
216 in.
228 in.
240 in.
252 in.
Unit Size:
Each
Product Code. 15957311
Brand: Metro™ TTS17NA
Only undefined left
Add to basket
Only undefined left
Add to basket
For use with Metro Super Erecta™, Super Erecta Pro™ and Super Adjustable™ Top Track™ shelving.
  • Track sets include necessary sections of track for assembling track runs up to 21 ft. (640 cm)
  • Coated with Microban™ antimicrobial protection
Order Info
19-138-228 (TTS2.5NA) includes a joining kit to connect the track to other track sets.
Specifications
Product Type Track Set
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ top track shelving
Includes Necessary sections of track for assembling track runs of 1830 to 6405mm
Length (English) 204 in.
Length (Metric) 518.2 cm
Product Content Correction
Product Title
Metro™ Wire Shelving Track Sets >