Läs mer
Metsulfuron-methyl, TRC
Produktkod. 30482796
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482796 250 mg
30470707 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30482796 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM227748250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74223-64-6
Molekylformel C14 H15 N5 O6 S
Molekylvikt (g/mol) 381.36
Synonym Benzoic acid, 2-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]-, methyl ester, Accurate 60VG, Algrip, Allie SX, Ally, Ally 20DF, Ally XP, Brush-off, Cimarron, DPD 63760H, DPX 6376, DPX-T 6376, Escort, Escort (pesticide), Grench, Gropper, HCHA 92HA, Laren, MSM Turf, Metsulfuron-methyl, N-[(2-Methoxycarbonyl)phenyl]sulfonyl-N'-(6-methoxy-4-methyl-2-triazinyl)urea, Saber DF, Strech 60VP, T 6376, Zinger, Zinger WP
IUPAC-namn methyl 2-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoate
LEDER COC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2
Specifikationer
Kemiskt namn eller material Metsulfuron-methyl
Kvantitet 250 mg
InChI formel InChI=1S/C14H15N5O6S/c1-8-15-12(18-14(16-8)25-3)17-13(21)19-26(22,23)10-7-5-4-6-9(10)11(20)24-2/h4-7H,1-3H3,(H2,15,16,17,18,19,21)
Formel vikt 381.0743