Läs mer
Metsulfuron-methyl, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Metsulfuron-methyl |
| CAS | 74223-64-6 |
| Molekylformel | C14 H15 N5 O6 S |
| Kvantitet | 1 g |
| Synonym | Benzoic acid, 2-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]-, methyl ester, Accurate 60VG, Algrip, Allie SX, Ally, Ally 20DF, Ally XP, Brush-off, Cimarron, DPD 63760H, DPX 6376, DPX-T 6376, Escort, Escort (pesticide), Grench, Gropper, HCHA 92HA, Laren, MSM Turf, Metsulfuron-methyl, N-[(2-Methoxycarbonyl)phenyl]sulfonyl-N'-(6-methoxy-4-methyl-2-triazinyl)urea, Saber DF, Strech 60VP, T 6376, Zinger, Zinger WP |
| InChI formel | InChI=1S/C14H15N5O6S/c1-8-15-12(18-14(16-8)25-3)17-13(21)19-26(22,23)10-7-5-4-6-9(10)11(20)24-2/h4-7H,1-3H3,(H2,15,16,17,18,19,21) |
| LEDER | COC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2 |
| IUPAC-namn | methyl 2-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoate |
| Molekylvikt (g/mol) | 381.36 |
| Formel vikt | 381.0743 |