Läs mer
Metsulfuron, TRC
Produktkod. 17972725
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17972725 10 mg
17953525 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17972725 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22774510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79510-48-8
Molekylformel C13 H13 N5 O6 S
Molekylvikt (g/mol) 367.34
Synonym 2-[[[[(4-Methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]benzoic acid (ACI), Cimarron 60DF, Escort XP, Metsulfuron, Benzoic acid, 2-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]- (9CI, ACI)
IUPAC-namn 2-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoic acid
LEDER COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2C(=O)O)n1
Specifikationer
Kemiskt namn eller material Metsulfuron
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C13H13N5O6S/c1-7-14-11(17-13(15-7)24-2)16-12(21)18-25(22,23)9-6-4-3-5-8(9)10(19)20/h3-6H,1-2H3,(H,19,20)(H2,14,15,16,17,18,21)
Formel vikt 367.0587
Procent renhet >95%