Learn More
Mettler Toledo™ Moisture Analyzer Sample Pans
Product Code. 15979405 Shop All Mettler Toledo Products
Change view
Click to view available options
Material:
Aluminum
For Use With (Equipment):
LJ16 Infrared Moisture Analyzer
Unit Size:
Pack of 80
Product Code. Material For Use With (Equipment) Unit Size
15979405 Aluminum LJ16 Infrared Moisture Analyzer Pack of 80
1 options
Product Code. 15979405 Supplier Mettler Toledo™ Supplier No. 13865
 Restricted Item
Beräknad leverans: 09-12-2026
Mettler Toledo sample pans are the only sample pans that are officially supported by Mettler Toledo Moisture Analyzers for optimal use and guaranteed performance.
Choice of washable, reusable stainless chrome-nickel steel pans for halogen moisture analyzers, or aluminum or stainless-steel pans for all Mettler Toledo moisture analyzers.
  • 3.75 in. (96mm) in diameter
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Bottom Shape Flat
Color Silver
Disposable Reusable
Material Aluminum
Shape Round
Sterility Non-sterile
Type Moisture Pan
For Use With (Equipment) LJ16 Infrared Moisture Analyzer
Quantity 80 Pk.
Diameter (Metric) 88.9 mm