Learn More
Mettler Toledo™ uMix™ Magnetic Stirrer
Provides an extra degree of precision for your applications.
Brand: Mettler Toledo™ 30040000
Additional Details : Weight : 0.60000kg
Description
Powered and controlled externally, the magnetic stirrer allows for enhanced reproducibility of results and higher efficiency in the measuring process.
The magnetic stirrer can be combined with either the SevenCompact™ pH/Ion S220, the SevenCompact Conductivity S230, or the multiparameter SevenExcellence™ benchtop meters.
Specifications
Magnetic stirrer uMix | |
Magnetic stirrer uMix |