Läs mer
Metyrapone, TRC
Produktkod. 30476928
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30476928 250 mg
1 options
Produktkod. 30476928 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM338990250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Metyrapone
CAS 54-36-4
Molekylformel C14 H14 N2 O
Kvantitet 250 mg
Synonym 1-Propanone, 2-methyl-1,2-di-3-pyridinyl-, 1-Propanone, 2-methyl-1,2-di-3-pyridyl- (6CI,8CI), 1,2-Bis(3-pyridyl)-2-methyl-1-propanone, 2-Methyl-1,2-bis(3-pyridyl)-1-propanone, 2-Methyl-1,2-di(β-pyridyl)-1-propanone, 2-Methyl-1,2-di-3-pyridyl-1-propanone, Mepyrapone, Methapyrapone, Methbipyranone, Methopirapone, Methopyrapone, Methopyrinine, Methopyrone, Metopiron, Metopirone, Metopyrone, Metroprione, Metyrapon, Metyrapone, NSC 25265, Su 4885
InChI formel InChI=1S/C14H14N2O/c1-14(2,12-6-4-8-16-10-12)13(17)11-5-3-7-15-9-11/h3-10H,1-2H3
LEDER CC(C)(C(=O)c1cccnc1)c2cccnc2
IUPAC-namn 2-methyl-1,2-dipyridin-3-ylpropan-1-one
Molekylvikt (g/mol) 226.27
Formel vikt 226.1106