Läs mer
Mevinphos, TRC
Produktkod. 30473603
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473603 100 mg
30479908 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30473603 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM340535100MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7786-34-7
Molekylformel C7 H13 O6 P
Molekylvikt (g/mol) 224.15
Synonym Crotonic acid, 3-hydroxy-, methyl ester, dimethyl phosphate (6CI,8CI), 1-Methoxycarbonyl-1-propen-2-yl dimethyl phosphate, 2-Carbomethoxy-1-methylvinyl dimethyl phosphate, 2-Methoxycarbonyl-1-methylvinyl dimethyl phosphate, CMDP, Fosdrin, Gesfid, Meniphos, Mevinphos, NSC 46470, O,O-Dimethyl O-(1-carbomethoxy-1-propen-2-yl) phosphate, PD 5, PD 5 (pesticide), Phosdrin, Phosdrin W 10
IUPAC-namn methyl 3-dimethoxyphosphoryloxybut-2-enoate
LEDER COC(=O)C=C(C)OP(=O)(OC)OC
Specifikationer
Kemiskt namn eller material Mevinphos
Kvantitet 100 mg
InChI formel InChI=1S/C7H13O6P/c1-6(5-7(8)10-2)13-14(9,11-3)12-4/h5H,1-4H3
Formel vikt 224.045