Läs mer
Mexiletine Hydrochloride, TRC
Produktkod. 30470392
Change view
Kvantitet:
50 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470392 1 g
30467000 50 mg
30475718 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30470392 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3408001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5370-01-4
Molekylformel C11 H17 N O . Cl H
Molekylvikt (g/mol) 215.72
Synonym Mexiletine Hydrochloride, 1-(2,6-Xylyloxy)-2-propylamine Hydrochloride, 1-(2',6'-Dimethylphenoxy)-2-aminopropane Hydrochloride, 1-Methyl-2-(2,6-xylyloxy)ethylamine Hydrochloride, Ko-1173
IUPAC-namn 1-(2,6-dimethylphenoxy)propan-2-amine;hydrochloride
LEDER Cl.CC(N)COc1c(C)cccc1C
Specifikationer
Kemiskt namn eller material Mexiletine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C11H17NO.ClH/c1-8-5-4-6-9(2)11(8)13-7-10(3)12;/h4-6,10H,7,12H2,1-3H3;1H
Formel vikt 215.1077
Procent renhet >95%