Läs mer
Mexrenone, TRC
Produktkod. 30480844
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30480844 10 mg
1 options
Produktkod. 30480844 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM34084010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7alpha-(Methoxycarbonyl)-3-oxo-17alpha-pregn-4-ene-21,17-carbolactone
CAS 41020-65-9
Molekylformel C24 H32 O5
Kvantitet 10 mg
Synonym Pregn-4-ene-7,21-dicarboxylic acid, 17-hydroxy-3-oxo-, γ-lactone, 7-methyl ester, (7α,17α)-, Pregn-4-ene-7,21-dicarboxylic acid, 17-hydroxy-3-oxo-, γ-lactone, methyl ester, (7α,17α)- (9CI), Spiro[17H-cyclopenta[a]phenanthrene-17,2'(5'H)-furan], pregn-4-ene-7,21-dicarboxylic acid deriv., 17-Hydroxy-3-oxo-17α-pregn-4-ene-7α,21-dicarboxylic acid methyl ester γ-lactone, 17-Hydroxy-7α-(methoxycarbonyl)-3-oxo-17α-pregn-4-ene-21-carboxylic acid lactone, 7α-(Methoxycarbonyl)canrenone, Mexrenone, SC 25152, ZK 32055, 7α-(Methoxycarbonyl)-3-oxo-17α-pregn-4-ene-21,17-carbolactone
InChI formel InChI=1S/C24H32O5/c1-22-8-4-15(25)12-14(22)13-16(21(27)28-3)20-17(22)5-9-23(2)18(20)6-10-24(23)11-7-19(26)29-24/h12,16-18,20H,4-11,13H2,1-3H3/t16-,17+,18+,20-,22+,23+,24-/m1/s1
LEDER COC(=O)[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]45CCC(=O)O5)[C@H]13
IUPAC-namn methyl (7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-carboxylate
Molekylvikt (g/mol) 400.5079
Formel vikt 400.225