Läs mer
Mezlocillin Sodium, TRC
Produktkod. 30009477
Change view
Kvantitet:
50 mg
100 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009477 5 g
30006924 50 mg
30003499 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009477 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3408505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 42057-22-7
Molekylformel C21 H24 N5 O8 S2 . Na
Molekylvikt (g/mol) 561.5637
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-6-[[(2R)-2-[[[3-(methylsulfonyl)-2-oxo-1-imidazolidinyl]carbonyl]amino]-2-phenylacetyl]amino]-7-oxo-, sodium salt (1:1), (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-6-[[(2R)-[[[3-(methylsulfonyl)-2-oxo-1-imidazolidinyl]carbonyl]amino]phenylacetyl]amino]-7-oxo-, monosodium salt, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-6-[[[[[3-(methylsulfonyl)-2-oxo-1-imidazolidinyl]carbonyl]amino]phenylacetyl]amino]-7-oxo-, monosodium salt, [2S-[2α,5α,6β(S*)]]-, (2S,5R,6R)-3,3-Dimethyl-6-[[(2R)-2-[[[3-(methylsulfonyl)-2-oxo-1-imidazolidinyl]carbonyl]amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Sodium Salt, (2S,5R,6R)-3,3-Dimethyl-6-[[(2R)-[[[3-(methylsulfonyl)-2-oxo-1-imidazolidinyl]carbonyl]amino]phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Monosodium Salt, [2S-[2α,5α,6β(S*)]]-3,3-Dimethyl-6-[[[[[3-(methylsulfonyl)-2-oxo-1-imidazolidinyl]carbonyl]amino]phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Monosodium Salt, Baycipen, Baypen, Mezlin, Mezlocillin sodium salt, Monosodium mezlocillin, Sodium mezlocillin
IUPAC-namn sodium;(2S,5R,6R)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
LEDER [Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](NC(=O)N3CCN(C3=O)S(=O)(=O)C)c4ccccc4)C(=O)N2[C@H]1C(=O)[O-]
Specifikationer
Kemiskt namn eller material Mezlocillin Sodium
Kvantitet 5 g
InChI formel InChI=1S/C21H25N5O8S2.Na/c1-21(2)14(18(29)30)26-16(28)13(17(26)35-21)22-15(27)12(11-7-5-4-6-8-11)23-19(31)24-9-10-25(20(24)32)36(3,33)34;/h4-8,12-14,17H,9-10H2,1-3H3,(H,22,27)(H,23,31)(H,29,30);/q;+1/p-1/t12-,13-,14+,17-;/m1./s1
Formel vikt 561.0964