missing translation for 'onlineSavingsMsg'
Läs mer

Novus Biologicals™ MFG-E8 Overexpression Lysate

Produktkod. 18282533 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet Unit Size
18282533 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18282533 Supplier Novus Biologicals™ Supplier No. NBL113037
  • ONLINE EXCLUSIVE PRICE
    7270.00 SEK
    6874.35 SEK / 0.1mg
    Save 395.65 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The MFG-E8 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Celllinje The lysate was created in HEK293T cells, using plasmid ID RC217163 and based on accession number NM_5928. The protein contains a C-terminal DDK tag.
För användning med (applikation) Western Blot
Kvantitet 0.1 mg
Förvaringsbuffert RIPA buffer
Renhet Protein
Testspecificitet Homo sapiens milk fat globule-EGF factor 8 protein (MFGE8), transcript variant 1, mRNA.
Innehåll och lagring -80°C. Avoid freeze-thaw cycles.
Proteintillstånd Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.