missing translation for 'onlineSavingsMsg'
Learn More

MGAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15103-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824124

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. UDP-N-acetylglucosamine:alpha-3-D-mannoside beta-1, 2-N-acetylglucosaminyltransferase I is a medial-Golgi enzyme essential for the synthesis of hybrid and complex N-glycans. The protein, encoded by a single exon, shows typical features of a type II transmembrane protein. The protein is believed to be essential for normal embryogenesis. Several variants encoding the same protein have been found for this gene.
Specifications

Specifications

MGAT1
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P26572, P27808, Q09325
GGNT1, GlcNAc T I, GLCNAC TI, GLCT1, GLYT1, GNT 1, GNT I, MGAT, MGAT1
Rabbit
IgG
150 μL
-20°C
Primary
17308, 4245, 81519
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MGAT1
Liquid
Mgat1
MGAT1 Fusion Protein Ag7446
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers