missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MGC26816 Recombinant Protein
Brand: Abnova™ H00164684-P01.10ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH22546 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
60.1 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MAVNQSHTENRRGALIPNGESLLKRSPNVELSFPQRSEGSNVFSGRKTGTLFLTSYRVIFITSCSISDPMLSFMMPFDLMTNLTVEQPVFAANFIKGTIQAAPYGGWEGQATFKLVFRNGGAIEFAQLMVKAASAAARGFPLRTLNDWFSSMGIYVITGEGNMCTPQMPCSVIVYGAPPAGYGAPPPGYGAPPAGYGAQPVGNEGPPVGYRASPVRYGAPPLGYGAPPAGYGAPPLGYGAPPLGYGTPPLGYGAPPLGYGAPPAGNEGPPAGYRASPAGSGARPHESTAAQAPENEASLPSASSSQVHS | |
FLJ26145/MGC26816/PAWP | |
WBP2NL | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, ELISA, Protein Array, Western Blot | |
164684 | |
MGC26816 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WBP2NL | |
Human | |
Recombinant | |
Solution |