missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGEA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | MGEA5 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823494
|
Proteintech
14711-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813494
|
Proteintech
14711-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Two bands at 130 kDa and 75 kDa could be detected using antibody 66033-1-Ig.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predomitly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.Specifications
MGEA5 | |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O60502, Q8VIJ5, Q9EQQ9 | |
10724, 154968, 76055 | |
MGEA5 Fusion Protein Ag6405 | |
Primary | |
-20°C | |
MGEA5 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Beta N acetylhexosaminidase, Bifunctional protein NCOAT, HAT, HEXC, Hexosaminidase C, KIAA0679, MEA5, Meningioma expressed antigen 5, MGEA5, N acetyl beta glucosaminidase, NCOAT, O GlcNAcase, OGA | |
MGEA5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |