missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549742
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total mGluR8 protein.
mGluR8 Polyclonal antibody specifically detects mGluR8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
mGluR8 | |
Polyclonal | |
Unconjugated | |
GRM8 | |
A230002O04; FLJ41058; G protein coupled receptor, family C, group 1, member H; G protein-coupled receptor family C group 1 member H; G protein-coupled receptor, family C, group 1, member H; GLUR8; glutamate metabotropic receptor 8; glutamate receptor; glutamate receptor, metabotropic 8; Gprc1h; Grm8; M mGlu8; Metabotropic glutamate receptor 8; metabotropic glutamate receptor subtype 8b; MGC126724; mGlu8; mGlur; mGluR8; mGluR8b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14823, 2918, 60590 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O00222, P47743, P70579 | |
GRM8 | |
Synthesized peptide derived from human GluR8. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |