missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGME1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23178-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody detects the 35-40 kDa protein.
The function of this protein remains unknown.Specifications
MGME1 | |
Polyclonal | |
Unconjugated | |
MGME1 | |
C20orf72, DDK1, MGME1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
74528, 92667 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BQP7, Q9CXC3 | |
Mgme1 | |
MGME1 Fusion Protein Ag19644 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |