missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGMT (O-methylguanine-DNA methyltransferase) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ MGMT (O-methylguanine-DNA methyltransferase) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
1259.70 SEK - 9605.00 SEK
Specifications
Antigen | MGMT (O-methylguanine-DNA methyltransferase) Ab-1 |
---|---|
Clone | MT 3.1 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12681527
|
Epredia
MS-470-B1 |
Concentrated | Biotin | 500 μL |
5990.00 SEK
500µL |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12651527
|
Epredia
MS-470-B |
Concentrated | Biotin | 1 mL |
9605.00 SEK
1mL |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12601537
|
Epredia
MS-470-P1 |
Concentrated | Unconjugated | 500 μL |
4595.00 SEK
500µL |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12671527
|
Epredia
MS-470-P |
Concentrated | Unconjugated | 1 mL |
7650.00 SEK
1mL |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12611537
|
Epredia
MS-470-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12621537
|
Epredia
MS-470-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12661527
|
Epredia
MS-470-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12691527
|
Epredia
MS-470-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651537
|
Epredia
MS-470-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Cancer chemotherapeutic alkylating agents (e.g. BCNU,) act by inducing formation of lethal cross-links at the O-alkylguanine position in DNA. MGMT transfers alkyl adducts from the O-position of guanine in DNA (prior to cross-link formation) to a cysteine residue in its own sequence, thereby restoring DNA to its intact state. This transfer inactivates the MGMT enzyme and is irreversible; hence the level of MGMT in a cell is directly proportional to the level of DNA-damage it can tolerate. In normal tissues, MGMT acts as a suppressor of mutation and carcinogenesis. Tumors with high levels of MGMT are likely be drug-resistant.Host Species: Mouse
Clone: MT 3.1
Isotype: IgG1
Species Reactivity: Human. Also reacts weakly with Mouse, Rat and Hamster. Others not known.
Epitope: aa 8-221
Immunogen: Purified recombinant human MGMT protein
Molecular Weight: 25kDa
Positive Control: Raji cells, Colon CA
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
MGMT (O-methylguanine-DNA methyltransferase) Ab-1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Enzyme | |
Purified recombinant human MGMT protein | |
Primary |
MT 3.1 | |
Monoclonal | |
RUO | |
Hamster, Human, Mouse, Rat | |
IgG1 |