Learn More
MGRN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552461
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LPFRALLQIR AVRKKPGALS PVSFSPVLAQ SLEHDEHSNS DSVPPGYEPI SLLEALNGLR AVSPAIPSAP LYEEITYSGI SDGLSQASCP LAAIDHILDS SRQKGRPQSK APDSTLRSPS SPIHEEDE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
MGRN1 Polyclonal antibody specifically detects MGRN1 in Human samples. It is validated for Immunohistochemistry.Specifications
MGRN1 | |
Polyclonal | |
Unconjugated | |
MGRN1 | |
2610042J20Rik; E3 ubiquitin-protein ligase MGRN1; KIAA0544; mahoganoid; mahogunin ring finger 1; mahogunin ring finger 1, E3 ubiquitin protein ligase; mahogunin RING finger protein 1; mahogunin, ring finger 1; mahogunin, ring finger 1a; md; mgrn1; mgrn1a; mKIAA0544; nc; probable E3 ubiquitin-protein ligase MGRN1; RGD1311862; RING finger protein 156; RING-type E3 ubiquitin transferase MGRN1; RNF156; wu:fi38c03; zgc:55978 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23295 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60291 | |
MGRN1 | |
Recombinant protein corresponding to Human MGRN1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only