Learn More
MGST1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545080
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: NPEDCVAFGK GENAKKYLRT DDRVERVRRA HLNDLENIIP FLGIGLLYSL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 86%; Rat: 93%
MGST1 Polyclonal antibody specifically detects MGST1 in Human samples. It is validated for Western Blot.Specifications
MGST1 | |
Polyclonal | |
Unconjugated | |
MGST1 | |
1500002K10Rik; glutathione S-transferase; glutathione S-transferase 12; Gst; GST12; MGC14525; MGST; mGST1; MGST-I; Microsomal glutathione S-transferase 1; microsomal glutathione S-transferase 1-like protein; microsomal glutathione-S-transferase; microsomal GST-1; Microsomal GST-I | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4257 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P10620 | |
MGST1 | |
Synthetic peptide directed towards the N-terminal of human MGST1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |