missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MHC Class II Polymorphic Antibody (CI.13.I), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64962
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MHC Class II Polymorphic Monoclonal specifically detects MHC Class II Polymorphic in Guinea Pig samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifikationer
| MHC Class II Polymorphic | |
| Monoclonal | |
| Unconjugated | |
| Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Reacts with the MHC Class II epitope present on strain 13, but not strain 2 guinea pig cells. The antigen is also found in outbred Hartley guinea pigs. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| CI.13.I | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 | |
| HLA-DRB1 | |
| Guinea pig peritoneal T-cells | |
| 0.125 mL | |
| Immunology, Major Histocompatibility Complexes | |
| 3123 | |
| Guinea Pig | |
| Supernatant |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering