missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC II (HLA-DR) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ MHC II (HLA-DR) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
1525.00 SEK - 5260.00 SEK
Specifications
Antigen | MHC II (HLA-DR) Ab-1 |
---|---|
Clone | LN3 |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12629616
|
Epredia
MS-133-P0 |
200μg/mL | 100 μL |
1525.00 SEK
100µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12639616
|
Epredia
MS-133-P1 |
200μg/mL | 500 μL |
3640.00 SEK
500µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12619616
|
Epredia
MS-133-P |
200μg/mL | 1 mL |
4680.00 SEK
1mL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12649616
|
Epredia
MS-133-P1ABX |
1mg/mL | 100 μL |
4240.00 SEK
100µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12659616
|
Epredia
MS-133-PABX |
1mg/mL | 200 μL |
5260.00 SEK
200µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of an α chain (36 kDa) and a β chain (27 kDa). It is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They therefore have a critical role in the initiation of the immune response.Host Species: Mouse
Clone: LN3
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Activated human peripheral blood mononuclear cells
Molecular Weight: 36kDa (α-chain) and 27kDa (β-chain)
Positive Control: Tonsil/lymph node
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
MHC II (HLA-DR) Ab-1 | |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Hematopoietic | |
Activated human peripheral blood mononuclear cells | |
Primary |
LN3 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG2b |