missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MI-463, MedChemExpress
Description
MI-463 is a highly potent and orally bioavailable small molecule inhibitor of the menin-mLL interaction.
Specifications
Specifications
| Kemiskt namn eller material | MI-463 |
| CAS | 1628317-18-9 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C24H23F3N6S |
| Kvantitet | 1 mg |
| Löslighetsinformation | DMSO : 125 mg/mL (257.98 mM; Need ultrasonic) |
| LEDER | FC(F)(F)CC(S1)=CC2=C1N=CN=C2NC3CCN(CC4=CC=C(NC(C#N)=C5)C5=C4C)CC3 |
| Molekylvikt (g/mol) | 484.54 |
| Formel vikt | 484.54 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?