Läs mer
Mibefradil Dihydrochloride, TRC
Produktkod. 30466204
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30466204 10 mg
1 options
Produktkod. 30466204 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM34180310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mibefradil Dihydrochloride
CAS 116666-63-8
Molekylformel C29H38FN3O3 . 2HCl
Kvantitet 10 mg
Synonym Acetic acid, 2-methoxy-, (1S,2S)-2-[2-[[3-(1H-benzimidazol-2-yl)propyl]methylamino]ethyl]-6-fluoro-1,2,3,4-tetrahydro-1-(1-methylethyl)-2-naphthalenyl ester, hydrochloride (1:2) (ACI), Acetic acid, methoxy-, (1S,2S)-2-[2-[[3-(1H-benzimidazol-2-yl)propyl]methylamino]ethyl]-6-fluoro-1,2,3,4-tetrahydro-1-(1-methylethyl)-2-naphthalenyl ester, dihydrochloride (9CI), Acetic acid, methoxy-, 2-[2-[[3-(1H-benzimidazol-2-yl)propyl]methylamino]ethyl]-6-fluoro-1,2,3,4-tetrahydro-1-(1-methylethyl)-2-naphthalenyl ester, dihydrochloride, (1S-cis)- (ZCI), Mibefradil dihydrochloride, Posicor, Ro 40-5967, Ro 40-5967/001, (1S,2S)-2-[2-[[3-(1H-Benzimidazol-2-yl)propyl]methylamino]ethyl]-6-fluoro-1-isopropyl-1,2,3,4-tetrahydronaphthalen-2-yl methoxyacetate dihydrochloride
InChI formel InChI=1S/C29H38FN3O3.2ClH/c1-20(2)28-23-12-11-22(30)18-21(23)13-14-29(28,36-27(34)19-35-4)15-17-33(3)16-7-10-26-31-24-8-5-6-9-25(24)32-26;;/h5-6,8-9,11-12,18,20,28H,7,10,13-17,19H2,1-4H3,(H,31,32);2*1H/t28-,29-;;/m0../s1
LEDER Cl.Cl.COCC(=O)O[C@]1(CCN(C)CCCc2nc3ccccc3[nH]2)CCc4cc(F)ccc4[C@@H]1C(C)C
IUPAC-namn [(1S,2S)-2-[2-[3-(1H-benzimidazol-2-yl)propyl-methylamino]ethyl]-6-fluoro-1-propan-2-yl-3,4-dihydro-1H-naphthalen-2-yl] 2-methoxyacetate;dihydrochloride
Molekylvikt (g/mol) 568.55
Formel vikt 567.2431