missing translation for 'onlineSavingsMsg'
Learn More

MICA/B Mouse anti-Human, Alexa Fluor® 488, Clone: 6D4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-5788-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15500227

  • 1290.00 SEK / 25 tests


EU Stock 3
Add to basket

Description

Description

Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. MICA encodes the higly polymorphic MHC (HLA) class I chain-related gene A. The protein product is expressed on the cell surface, although unlike canonical class I molecules does not seem to associate with beta-2-microglobulin. It is thought that MICA functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells.
Specifications

Specifications

MICA/B
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q29980, Q29983
MHC class I polypeptide-related sequence A, MHC class I polypeptide-related sequence B
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
6D4
Alexa Fluor® 488
MICA
Liquid
MICA, MICB
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
100507436, 4277
Videos

For Research Use Only.