Läs mer
Micafungin Sodium, TRC
Produktkod. 17950555
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17950555 100 mg
17941475 5 mg
17915075 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17950555 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM342350100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 208538-73-2
Synonym 1-[(4R,5R)-4,5-Dihydroxy-N2-[4-[5-[4-(pentyloxy)phenyl]-3-isoxazolyl]benzoyl]-L-ornithine]-4-[(4S)-4-hydroxy-4-[4-hydroxy-3-(sulfooxy)phenyl]-L-threonine]pneumocandin A0;
Specifikationer
Renhet >95% (HPLC)
Formatera Neat
Molekylvikt (g/mol) 1292.26
Produkttyp API