missing translation for 'onlineSavingsMsg'
Learn More

MICB Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14325-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882904

  • 3248.87 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MICA encodes the higly polymorphic MHC (HLA) class I chain-related gene A. The protein product is expressed on the cell surface, although unlike canonical class I molecules does not seem to associate with beta-2-microglobulin. It is thought that MICA functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells.
Specifications

Specifications

MICB
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q29980
MIC B, MICB, PERB11.2
Rabbit
IgG
150 μL
-20°C
Primary
4277
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MICB
Liquid
MICB
MICB Fusion Protein Ag5591
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers