Läs mer
Miconazole EP Impurity F (as Nitrate), Mikromol™
Beskrivning
Miconazole EP Impurity F (as Nitrate)
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-[(2RS)-2-[(3,4-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate |
| CAS | 27220-35-5 |
| Analyt- eller komponentnamn | 1-[(2RS)-2-[(3,4-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate |
| Frakt skick | Room Temperature |
| Molekylformel | C18 H14 Cl4 N2 O . H N O3 |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Miconazole Nitrate Imp. F (EP), Miconazole USP Related Compound F, Miconazole USP RC F, 1-[(2RS)-2-[(3,4-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. F (EP) as Nitrate, Miconazole Related Compound F as Nitrate, Miconazole Impurity F as Nitrate, Miconazole Nitrate Impurity F as Nitrate, Miconazole Impurity F as Nitrate |
| InChI formel | InChI=1S/C18H14Cl4N2O.HNO3/c19-13-2-3-14(16(21)8-13)18(9-24-6-5-23-11-24)25-10-12-1-4-15(20)17(22)7-12;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4) |
| LEDER | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccc(Cl)c(Cl)c3)c(Cl)c1 |
| IUPAC-namn | 1-[2-(2,4-dichlorophenyl)-2-[(3,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid |
| Visa mer |