Läs mer
Miconazole Impurity B, TRC
Produktkod. 30480739
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480739 5 mg
30465891 50 mg
30465379 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480739 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3424855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 913837-72-6
Molekylformel C18H15Cl3N2O
Molekylvikt (g/mol) 381.68
Synonym 1-[2-[(3-Chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]1H-Imidazole
IUPAC-namn 1-[2-[(3-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole
LEDER Clc1cccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c1
Specifikationer
Kemiskt namn eller material Miconazole Impurity B
Kvantitet 5 mg
InChI formel InChI=1S/C18H15Cl3N2O/c19-14-3-1-2-13(8-14)11-24-18(10-23-7-6-22-12-23)16-5-4-15(20)9-17(16)21/h1-9,12,18H,10-11H2
Formel vikt 380.025