Läs mer
Miconazole Impurity H, TRC
Produktkod. 30134160
Change view
Kvantitet:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134160 100 mg
30133561 1 g
30004440 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134160 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM342490100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 181931-30-6
Molekylformel C18H16Cl2N2O
Molekylvikt (g/mol) 347.24
Synonym 1-[2-(2,4-Dichlorophenyl)-2-(phenylmethoxy)ethyl]-1H-Imidazole, Miconazole EP Impurity H
LEDER ClC(C=C(Cl)C=C1)=C1C(CN2C=CN=C2)OCC3=CC=CC=C3
Specifikationer
Kemiskt namn eller material Miconazole Impurity H
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C18H16Cl2N2O/c19-15-6-7-16(17(20)10-15)18(11-22-9-8-21-13-22)23-12-14-4-2-1-3-5-14/h1-10,13,18H,11-12H2
Procent renhet >95%