Learn More
Uvex Micro Black/Orange PU/PU 65002 Sandal - Width 11
Optimal shock absorption, ergonomic fit, low weight and excellent climate characteristics ensure max performance at work and redefine the limits for safety shoes; range in protection classes S3 and S1 P
1575.00 SEK
Specifications
Color | Black, Orange |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17220571
|
Uvex
6500235 |
35 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17230571
|
Uvex
6500236 |
36 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17240571
|
Uvex
6500237 |
37 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17250571
|
Uvex
6500238 |
38 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17260571
|
Uvex
6500239 |
39 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17270571
|
Uvex
6500240 |
40 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17280571
|
Uvex
6500241 |
41 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17290571
|
Uvex
6500242 |
42 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17200581
|
Uvex
6500243 |
43 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17210581
|
Uvex
6500244 |
44 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17220581
|
Uvex
6500245 |
45 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17230581
|
Uvex
6500246 |
46 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17240581
|
Uvex
6500247 |
47 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17250581
|
Uvex
6500248 |
48 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17260581
|
Uvex
6500249 |
49 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17270581
|
Uvex
6500250 |
50 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17280581
|
Uvex
6500251 |
51 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17290581
|
Uvex
6500252 |
52 |
1575.00 SEK
1 pair |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Sandal 6500.2 S1 P SRC
- Sporty, multifunctional safety sandal
- Extremely light and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side and ankle twist stability, no thermal conductivity and anti-magnetic properties
- Metal-free, penetration-resistance insole in accordance with latest standard, which does not impact shoe flexibility
- Made with new high-performance technology for exceptional wearer comfort
- Uvex Climazone technology for optimum comfort
- Breathable, water-repellent, perforated high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic textiles, suitable for those allergic to chrome
- Two Velcro™ straps for individual fit
- Virtually seam-free construction to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue
- Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
- Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Uvex topshield scuffcap to protect the upper and extend its life
- Meets ESD specification with volume resistance under 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6500.2
- Standard EN ISO 20345:2011 S1 P SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Standard width | |
Yes | |
S1 |